ENA388191842 Enamine

4-(2,5-difluorophenyl)-4-oxobutanoic acid

  • MDL number MFCD01320107
  • Cas Number : 871127-78-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120598

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120598
Availability details for
Quantity
  • Availability
  • Add to Cart