ENA388191843 Enamine

4-chlorobenzene-1,3-disulfonamide

  • MDL number MFCD00210257
  • Cas Number : 671-95-4
  • Molecular Weight : 270.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120606

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120606
Availability details for
Quantity
  • Availability
  • Add to Cart