ENA388191845 Enamine

2-(3-phenyl-1H-1,2,4-triazol-1-yl)acetic acid

  • MDL number MFCD21882798
  • Cas Number : 1368811-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...181
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120626

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120626
Availability details for
Quantity
  • Availability
  • Add to Cart