ENA388191852 Enamine

1-[1-(2-methoxyethyl)-3,5-dimethyl-1H-pyrazol-4-yl]ethan-1-one

  • MDL number MFCD16861486
  • Cas Number : 1342025-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 31...33
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111727

Safety Information

msds https://www.enaminestore.com/catalog/EN300-111727
Availability details for
Quantity
  • Availability
  • Add to Cart