ENA388191857 Enamine

2-amino-3,3,3-trifluoro-2-(3-methoxyphenyl)propanoic acid

  • MDL number MFCD19065313
  • Cas Number : 1393585-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120687

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120687
Availability details for
Quantity
  • Availability
  • Add to Cart