ENA388191859 Enamine

5-chloro-2-(2-methoxyethoxy)benzene-1-sulfonyl chloride

  • MDL number MFCD16681756
  • Cas Number : 111762-21-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 38...40
Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120710

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120710
Availability details for
Quantity
  • Availability
  • Add to Cart