ENA388191864 Enamine

3-amino-1-ethyl-1,4-dihydropyridin-4-one

  • MDL number MFCD22741181
  • Cas Number : 1423025-23-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113056

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113056
Availability details for
Quantity
  • Availability
  • Add to Cart