ENA388191865 Enamine

methyl 4-(1H-pyrrol-1-yl)butanoate

  • MDL number MFCD22741182
  • Cas Number : 204767-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113354

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113354
Availability details for
Quantity
  • Availability
  • Add to Cart