ENA388191876 Enamine

ethyl 2-acetyl-4-oxopentanoate

  • MDL number MFCD00204229
  • Cas Number : 41892-81-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120816

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120816
Availability details for
Quantity
  • Availability
  • Add to Cart