ENA388191878 Enamine

2-(pyridin-3-yl)-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD07375357
  • Cas Number : 248275-42-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120841

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120841
Availability details for
Quantity
  • Availability
  • Add to Cart