ENA388191880 Enamine

1-(4,5,6,7-tetrahydro-1,3-benzothiazol-2-yl)ethan-1-one

  • MDL number MFCD18343967
  • Cas Number : 1211513-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120844

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120844
Availability details for
Quantity
  • Availability
  • Add to Cart