ENA388191882 Enamine

5H,6H,7H-pyrrolo[1,2-a]imidazole-3-sulfonyl chloride

  • MDL number MFCD08235266
  • Cas Number : 914637-94-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120848

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-120848
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart