ENA388191898 Enamine

methyl 2-amino-5-(trifluoromethyl)benzoate

  • MDL number MFCD08234902
  • Cas Number : 117324-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120884

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-120884
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart