ENA388191907 Enamine

[1-(2,4-difluorophenyl)cyclopropyl]methanamine

  • MDL number MFCD20362006
  • Cas Number : 1340550-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121007

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121007
Availability details for
Quantity
  • Availability
  • Add to Cart