ENA388191908 Enamine

2-amino-5-(2,3-dihydro-1H-indol-1-yl)benzoic acid

  • MDL number MFCD12845815
  • Cas Number : 1178442-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189...191
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121008

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121008
Availability details for
Quantity
  • Availability
  • Add to Cart