ENA388191912 Enamine

3-ethylpyridine-2-carbonitrile

  • MDL number MFCD05982125
  • Cas Number : 58515-54-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121013

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121013
Availability details for
Quantity
  • Availability
  • Add to Cart