ENA388191913 Enamine

2-(2-phenylethyl)-1,3-thiazolidine-4-carboxylic acid

  • MDL number MFCD01622693
  • Cas Number : 65469-04-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121014

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121014
Availability details for
Quantity
  • Availability
  • Add to Cart