ENA388191917 Enamine

N-methyl-1,2,3,4-tetrahydroquinolin-5-amine

  • MDL number MFCD22741234
  • Cas Number : 1423033-78-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121025

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-121025
Availability details for
Quantity
  • Availability
  • Add to Cart