ENA388191920 Enamine

4-ethyloxan-4-amine hydrochloride

  • MDL number MFCD22418725
  • Cas Number : 1385696-70-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121043

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-121043
Availability details for
Quantity
  • Availability
  • Add to Cart