ENA388191928 Enamine

1-bromo-3-(bromomethyl)-2-methylbenzene

  • MDL number MFCD11847532
  • Cas Number : 112299-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121065

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-121065
Availability details for
Quantity
  • Availability
  • Add to Cart