ENA388191931 Enamine

4-oxaspiro[bicyclo[3.2.0]heptane-6,1'-cyclobutane]-7-ol

  • MDL number MFCD22741187
  • Cas Number : 1423028-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115999

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115999
Availability details for
Quantity
  • Availability
  • Add to Cart