ENA388191934 Enamine

7-bromo-2,3,4,5-tetrahydro-1H-1-benzazepin-2-one

  • MDL number MFCD07371648
  • Cas Number : 53841-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119...121
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121084

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121084
Availability details for
Quantity
  • Availability
  • Add to Cart