ENA388191935 Enamine

(2-aminoethyl)(pyridin-2-ylmethyl)amine

  • MDL number MFCD04355991
  • Cas Number : 20947-95-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121085

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121085
Availability details for
Quantity
  • Availability
  • Add to Cart