ENA388191942 Enamine

2-[(3-methyl-1H-pyrazol-5-yl)carbamoyl]cyclohexane-1-carboxylic acid

  • MDL number MFCD19192459

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114...116
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121095

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121095
Availability details for
Quantity
  • Availability
  • Add to Cart