ENA388191953 Enamine

1-bromohex-5-en-2-one

  • MDL number MFCD20622719
  • Cas Number : 145967-22-2
  • Molecular Weight : 177.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-bromo-5-hexen-2-one
Inchi Code 1S/C6H9BrO/c1-2-3-4-6(8)5-7/h2H,1,3-5H2
Inchi Key SICVUWZDYJMQCT-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-121108

Safety Information

signalword Danger
pcodes P210,P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P310,P312,P332+P313,P362,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-121108
pictograms GHS05,GHS07
hstatements H227,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart