ENA388191965 Enamine

3-(ethanesulfonyl)benzoic acid

  • MDL number MFCD10032482
  • Cas Number : 861079-51-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160...162
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121246

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-121246
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart