ENA388191967 Enamine

7-methylimidazo[1,2-a]pyridine-3-carboxylic acid

  • MDL number MFCD01720461
  • Cas Number : 21801-80-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121248

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121248
Availability details for
Quantity
  • Availability
  • Add to Cart