ENA388191968 Enamine

5-(4-phenyl-4H-1,2,4-triazol-3-yl)pyridin-2-amine

  • MDL number MFCD18554229
  • Cas Number : 1423033-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121249

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-121249
Availability details for
Quantity
  • Availability
  • Add to Cart