ENA388191973 Enamine

4-(4-methyl-4H-1,2,4-triazol-3-yl)pyridin-2-amine

  • MDL number MFCD20307427
  • Cas Number : 1339170-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121262

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-121262
Availability details for
Quantity
  • Availability
  • Add to Cart