ENA388191983 Enamine

5-aminohexanoic acid hydrochloride

  • MDL number MFCD22741247
  • Cas Number : 855913-65-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121299

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-121299
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart