ENA388191993 Enamine

(5-methyl-4-propyl-4H-1,2,4-triazol-3-yl)methanol

  • MDL number MFCD22741253
  • Cas Number : 1423033-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59...61
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121534

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121534
Availability details for
Quantity
  • Availability
  • Add to Cart