ENA388191994 Enamine

4-(1H-pyrrol-2-yl)aniline

  • MDL number MFCD02823695
  • Cas Number : 134703-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121535

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-121535
Availability details for
Quantity
  • Availability
  • Add to Cart