ENA388191995 Enamine

3-methyl-4-oxocyclohexane-1-carboxylic acid

  • MDL number MFCD20638867
  • Cas Number : 101567-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117027

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117027
Availability details for
Quantity
  • Availability
  • Add to Cart