ENA388192001 Enamine

1-phenyl-N-(1H-pyrazol-4-yl)methanesulfonamide

  • MDL number MFCD12133976
  • Cas Number : 1156354-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121561

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121561
Availability details for
Quantity
  • Availability
  • Add to Cart