ENA388192002 Enamine

ethyl 2-methylpyrimidine-4-carboxylate

  • MDL number MFCD11045650
  • Cas Number : 76240-14-7
  • Molecular Weight : 166.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-121562

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-121562
Availability details for
Quantity
  • Availability
  • Add to Cart