ENA388192003 Enamine

2-{[4-amino-5-(4-methoxyphenyl)-4H-1,2,4-triazol-3-yl]sulfanyl}acetic acid

  • MDL number MFCD01166520
  • Cas Number : 305336-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-121563

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-121563
Availability details for
Quantity
  • Availability
  • Add to Cart