ENA388192004 Enamine

N-methyl-N-(pyridin-3-yl)acetamide

  • MDL number MFCD01566849
  • Cas Number : 55899-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121566

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-121566
Availability details for
Quantity
  • Availability
  • Add to Cart