ENA388192005 Enamine

1-(4-chlorophenyl)-3,3,3-trifluoropropan-1-amine

  • MDL number MFCD09809859
  • Cas Number : 1016531-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121568

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121568
Availability details for
Quantity
  • Availability
  • Add to Cart