ENA388192010 Enamine

3,5-dibromo-1H-pyrazole

  • MDL number MFCD00159645
  • Cas Number : 67460-86-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117520

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117520
Availability details for
Quantity
  • Availability
  • Add to Cart