ENA388192014 Enamine

4-bromo-3-(chlorosulfonyl)-5-methylbenzoic acid

  • MDL number MFCD09029773
  • Cas Number : 849333-58-0
  • Molecular Weight : 313.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-121581

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-121581
Availability details for
Quantity
  • Availability
  • Add to Cart