ENA388192017 Enamine

N-ethyl-3-fluorobenzene-1-sulfonamide

  • MDL number MFCD06680332
  • Cas Number : 852860-28-7
  • Molecular Weight : 203.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-ethyl-3-fluorobenzenesulfonamide
Inchi Code 1S/C8H10FNO2S/c1-2-10-13(11,12)8-5-3-4-7(9)6-8/h3-6,10H,2H2,1H3
Inchi Key ACWVKIUBBLNBHY-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-121587

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-121587
Availability details for
Quantity
  • Availability
  • Add to Cart