ENA388192024 Enamine

4-{[3-(methylamino)propyl]amino}butanoic acid dihydrochloride

  • MDL number MFCD22741264
  • Cas Number : 1423028-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99...101
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121802

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121802
Availability details for
Quantity
  • Availability
  • Add to Cart