ENA388192026 Enamine

3-(1-methyl-1H-pyrazol-4-yl)benzaldehyde

  • MDL number MFCD08669917
  • Cas Number : 179055-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121805

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121805
Availability details for
Quantity
  • Availability
  • Add to Cart