ENA388192027 Enamine

1,2,5-trimethyl-1H-indole-3-carbaldehyde

  • MDL number MFCD07364558
  • Cas Number : 66727-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104...106
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121807

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121807
Availability details for
Quantity
  • Availability
  • Add to Cart