ENA388192028 Enamine

2-[(3-methoxy-3-oxopropyl)sulfanyl]-5-nitrobenzoic acid

  • MDL number MFCD12487894
  • Cas Number : 1153234-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121808

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-121808
Availability details for
Quantity
  • Availability
  • Add to Cart