ENA388192031 Enamine

[2,3'-bipyridin]-6'-amine

  • MDL number MFCD12033340
  • Cas Number : 31860-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121812

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-121812
Availability details for
Quantity
  • Availability
  • Add to Cart