ENA388192033 Enamine

1-(3-chloropyridin-2-yl)-1H-pyrazol-4-amine

  • MDL number MFCD14594691
  • Cas Number : 1250279-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-121814

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-121814
Availability details for
Quantity
  • Availability
  • Add to Cart