ENA388192042 Enamine

3-{benzyl[2-(cyclopropylamino)ethyl]amino}propanoic acid dihydrochloride

  • MDL number MFCD22741270
  • Cas Number : 1423029-66-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121826

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121826
Availability details for
Quantity
  • Availability
  • Add to Cart