ENA388192047 Enamine

3-bromo-1-(4-chloro-3-fluorophenyl)pyrrolidin-2-one

  • MDL number MFCD18886840
  • Cas Number : 1339032-58-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94...96
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121863

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121863
Availability details for
Quantity
  • Availability
  • Add to Cart