ENA388192051 Enamine

3-amino-1-(4-chloro-3-fluorophenyl)pyrrolidin-2-one

  • MDL number MFCD14689913
  • Cas Number : 1251344-72-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69...71
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121867

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121867
Availability details for
Quantity
  • Availability
  • Add to Cart